Product Details
- SNP ID
-
rs28715769
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:75557029 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTCCCCGTTCCCATCCCTCAGAG[C/T]TCACAATAGACAGGAGGAGGGGAGA
- Phenotype
-
MIM: 614829
MIM: 612535
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C4orf26
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116623474] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4orf26
- Gene Name
- chromosome 4 open reading frame 26
- Gene
- THAP6
- Gene Name
- THAP domain containing 6
There are no transcripts associated with this gene.
View Full Product Details