Product Details

SNP ID
rs35101723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89726035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATGATTTTTATTATTAAAGTGA[G/T]ATGGGATAAAAATAAAATATTTTTG
Phenotype
MIM: 163890
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNCA PubMed Links

Gene Details

Gene
SNCA
Gene Name
synuclein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000345.3 1086 UTR 3 NP_000336.1
NM_001146054.1 1086 UTR 3 NP_001139526.1
NM_001146055.1 1086 UTR 3 NP_001139527.1
NM_007308.2 1086 UTR 3 NP_009292.1
XM_011532203.1 1086 Intron XP_011530505.1
XM_011532204.2 1086 Intron XP_011530506.1
XM_011532205.2 1086 Intron XP_011530507.1
XM_011532206.1 1086 Intron XP_011530508.1
XM_011532207.1 1086 Intron XP_011530509.1
XM_011532208.2 1086 UTR 3 XP_011530510.1
XM_017008562.1 1086 UTR 3 XP_016864051.1
XM_017008563.1 1086 UTR 3 XP_016864052.1

View Full Product Details