Product Details
- SNP ID
-
rs28635799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:80327890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGTTTAGCCAAGGTGGTGGAAT[A/T]AAAAAGTTACACTAATAAGAACAGG
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C4orf22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76828977] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4orf22
- Gene Name
- chromosome 4 open reading frame 22
View Full Product Details