Product Details

SNP ID
rs35773588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:145100975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGATGGATGGAGTACATTCTAC[C/T]TGGGGAGGGGAAAAATGTGTATGTG
Phenotype
MIM: 601213 MIM: 613745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCE1 PubMed Links

Gene Details

Gene
ABCE1
Gene Name
ATP binding cassette subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040876.1 Intron NP_001035809.1
NM_002940.2 Intron NP_002931.2
Gene
ANAPC10
Gene Name
anaphase promoting complex subunit 10
There are no transcripts associated with this gene.

View Full Product Details