Product Details

SNP ID
rs1803342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:91146242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTACAGAGATATTGTTGTTATGA[A/G]GGTAGACAACCTACAACATGAAACG
Phenotype
MIM: 125255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCN PubMed Links

Gene Details

Gene
DCN
Gene Name
decorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001920.4 1126 Missense Mutation CCT,CTT P299L NP_001911.1
NM_133503.3 1126 Missense Mutation CCT,CTT P299L NP_598010.1
NM_133504.3 1126 Missense Mutation CCT,CTT P190L NP_598011.1
NM_133505.3 1126 Missense Mutation CCT,CTT P152L NP_598012.1
NM_133506.3 1126 Missense Mutation CCT,CTT P112L NP_598013.1
NM_133507.3 1126 Silent Mutation CCC,CCT P74P NP_598014.1
XM_005268693.1 1126 Missense Mutation CCT,CTT P299L XP_005268750.1
XM_006719270.1 1126 Missense Mutation CCT,CTT P299L XP_006719333.1
XM_017018917.1 1126 Missense Mutation CCT,CTT P299L XP_016874406.1

View Full Product Details