Product Details

SNP ID
rs34169771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38068976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCTTTTTTAAAATTTTGGTAC[A/G]AAGATTCCAAAAAGCTTAATAAAAT
Phenotype
MIM: 601771 MIM: 611872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP1B1 PubMed Links

Gene Details

Gene
CYP1B1
Gene Name
cytochrome P450 family 1 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000104.3 3781 UTR 3 NP_000095.2
Gene
RMDN2
Gene Name
regulator of microtubule dynamics 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170791.2 3781 Intron NP_001164262.1
NM_001170792.2 3781 Intron NP_001164263.1
NM_001170793.2 3781 Intron NP_001164264.1
NM_001322211.1 3781 Intron NP_001309140.1
NM_001322212.1 3781 Intron NP_001309141.1
NM_144713.4 3781 Intron NP_653314.3
XM_011532613.2 3781 Intron XP_011530915.1
XM_011532614.2 3781 Intron XP_011530916.1
XM_011532615.2 3781 Intron XP_011530917.2
XM_011532616.2 3781 Intron XP_011530918.1
XM_011532617.2 3781 Intron XP_011530919.1
XM_011532618.2 3781 Intron XP_011530920.1
XM_011532619.2 3781 Intron XP_011530921.1
XM_017003474.1 3781 Intron XP_016858963.1
XM_017003475.1 3781 Intron XP_016858964.1
XM_017003476.1 3781 Intron XP_016858965.1
XM_017003477.1 3781 Intron XP_016858966.1
XM_017003478.1 3781 Intron XP_016858967.1
XM_017003479.1 3781 Intron XP_016858968.1

View Full Product Details