Product Details
- SNP ID
-
hCV62312841
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:75650162 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTGCAGGTGTGATTCAGACGTA[C/T]ACCTCTATGTATGTTGCAGTGAGCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FRG2C
PubMed Links
Gene Details
- Gene
- FRG2C
- Gene Name
- FSHD region gene 2 family member C
There are no transcripts associated with this gene.
View Full Product Details