Product Details

SNP ID
rs28362643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39107998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCGGTCCTGCCTTTTCTTGCC[C/T]CACTCCCCCTGGCAAGGGCGTCTTC
Phenotype
MIM: 606867 MIM: 611430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GORASP1 PubMed Links

Gene Details

Gene
GORASP1
Gene Name
golgi reassembly stacking protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278789.1 366 Intron NP_001265718.1
NM_001278790.1 366 Intron NP_001265719.1
NM_031899.3 366 UTR 5 NP_114105.1
XM_006713301.3 366 Intron XP_006713364.1
XM_011534020.2 366 Intron XP_011532322.1
XM_011534021.2 366 Intron XP_011532323.1
XM_017007050.1 366 Intron XP_016862539.1
XM_017007051.1 366 UTR 5 XP_016862540.1
XM_017007052.1 366 Intron XP_016862541.1
Gene
TTC21A
Gene Name
tetratricopeptide repeat domain 21A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105513.2 366 Intron NP_001098983.2
NM_145755.2 366 Intron NP_665698.2
XM_005264921.4 366 Intron XP_005264978.1
XM_005264922.4 366 Intron XP_005264979.1
XM_005264923.3 366 Intron XP_005264980.1
XM_005264924.4 366 Intron XP_005264981.1
XM_005264925.4 366 Intron XP_005264982.1
XM_005264926.4 366 Intron XP_005264983.1
XM_005264927.4 366 Intron XP_005264984.1
XM_006713011.3 366 Intron XP_006713074.1
XM_011533447.2 366 Intron XP_011531749.1
XM_011533449.1 366 Intron XP_011531751.1
XM_017005843.1 366 Intron XP_016861332.1

View Full Product Details