Product Details

SNP ID
rs28380655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89360916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTCAAGTATGTAATTATTTTG[A/T]TAAGTGTGAAAGTGCTTGAAATCCA
Phenotype
MIM: 607693 MIM: 601866
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SECISBP2 PubMed Links

Gene Details

Gene
SECISBP2
Gene Name
SECIS binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282688.1 2844 Intron NP_001269617.1
NM_001282689.1 2844 Intron NP_001269618.1
NM_001282690.1 2844 Intron NP_001269619.1
NM_024077.4 2844 Intron NP_076982.3
XM_005252196.2 2844 Intron XP_005252253.1
XM_005252202.3 2844 Intron XP_005252259.1
XM_006717282.2 2844 Intron XP_006717345.1
XM_011519000.2 2844 UTR 3 XP_011517302.1
XM_011519001.1 2844 Intron XP_011517303.1
XM_011519002.1 2844 Intron XP_011517304.1
XM_011519003.1 2844 Intron XP_011517305.1
XM_017015122.1 2844 Intron XP_016870611.1
XM_017015123.1 2844 Intron XP_016870612.1
XM_017015124.1 2844 Intron XP_016870613.1
XM_017015125.1 2844 Intron XP_016870614.1
Gene
SEMA4D
Gene Name
semaphorin 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142287.1 2844 UTR 3 NP_001135759.1
NM_006378.3 2844 Intron NP_006369.3
XM_005251654.3 2844 Intron XP_005251711.1
XM_011518123.2 2844 Intron XP_011516425.1
XM_011518124.2 2844 Intron XP_011516426.1
XM_011518125.1 2844 Intron XP_011516427.1
XM_011518127.2 2844 Intron XP_011516429.1
XM_011518128.2 2844 Intron XP_011516430.1
XM_011518129.1 2844 Intron XP_011516431.1
XM_011518130.2 2844 Intron XP_011516432.1
XM_011518131.2 2844 Intron XP_011516433.1
XM_011518133.2 2844 Intron XP_011516435.1
XM_011518134.2 2844 Intron XP_011516436.1
XM_017014193.1 2844 Intron XP_016869682.1
XM_017014194.1 2844 Intron XP_016869683.1
XM_017014195.1 2844 Intron XP_016869684.1
XM_017014196.1 2844 Intron XP_016869685.1
XM_017014197.1 2844 Intron XP_016869686.1
XM_017014198.1 2844 Intron XP_016869687.1
XM_017014199.1 2844 Intron XP_016869688.1

View Full Product Details