Product Details

SNP ID
rs28469288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150375694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGAGGATGGCCTCACAGGAAG[G/T]GGGATGACTGAGAGGCCCAGGTAGG
Phenotype
MIM: 612025
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IYD PubMed Links

Gene Details

Gene
IYD
Gene Name
iodotyrosine deiodinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164694.1 Intron NP_001158166.1
NM_001164695.1 Intron NP_001158167.1
NM_001318495.1 Intron NP_001305424.1
NM_203395.2 Intron NP_981932.1
XM_006715478.3 Intron XP_006715541.1
XM_006715479.3 Intron XP_006715542.1

View Full Product Details