Product Details

SNP ID
rs28597788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112382341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGATTTCTTGACCTCATGGTCC[A/G]CCCGCCTCAGCCTCCCAAAGTGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPACA7 PubMed Links

Gene Details

Gene
SPACA7
Gene Name
sperm acrosome associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145248.4 Intron NP_660291.2
XM_011537466.1 Intron XP_011535768.1
XM_011537467.2 Intron XP_011535769.1
XM_011537468.2 Intron XP_011535770.1
XM_011537471.2 Intron XP_011535773.1
XM_011537472.2 Intron XP_011535774.1
XM_017020391.1 Intron XP_016875880.1
XM_017020392.1 Intron XP_016875881.1

View Full Product Details