Product Details

SNP ID
rs17490613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:40226553 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGTCTATCTTTATCTTGATGTT[G/T]ATCTCTGTATCTAGCTGTATCTAGT
Phenotype
MIM: 609007
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRK2 PubMed Links

Gene Details

Gene
LRRK2
Gene Name
leucine rich repeat kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198578.3 Intron NP_940980.3
XM_005268629.3 Intron XP_005268686.1
XM_011537877.2 Intron XP_011536179.1
XM_011537881.2 Intron XP_011536183.1
XM_011537882.2 Intron XP_011536184.1
XM_017018786.1 Intron XP_016874275.1
XM_017018787.1 Intron XP_016874276.1
XM_017018788.1 Intron XP_016874277.1
XM_017018789.1 Intron XP_016874278.1

View Full Product Details