Product Details

SNP ID
rs1071634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101770341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGCAGCACCTATCGGGGCTC[C/G]GGGGCCCAAGTCCTCTGGCTGCCGC
Phenotype
MIM: 600483
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGF8 PubMed Links

Gene Details

Gene
FGF8
Gene Name
fibroblast growth factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206389.1 825 Silent Mutation CCC,CCG P137P NP_001193318.1
NM_006119.4 825 Silent Mutation CCC,CCG P212P NP_006110.1
NM_033163.3 825 Silent Mutation CCC,CCG P241P NP_149353.1
NM_033164.3 825 Silent Mutation CCC,CCG P230P NP_149354.1
NM_033165.3 825 Silent Mutation CCC,CCG P201P NP_149355.1

View Full Product Details