Product Details

SNP ID
rs17879858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79939321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCTGCTCAGAATGCAGGCTGC[A/C]CCCTGACTCATCGTCCTTAAGATTC
Phenotype
MIM: 178635
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BMS1P21 PubMed Links

Gene Details

Gene
BMS1P21
Gene Name
BMS1, ribosome biogenesis factor pseudogene 21
There are no transcripts associated with this gene.

Gene
SFTPD
Gene Name
surfactant protein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003019.4 Intron NP_003010.4
XM_011540087.1 Intron XP_011538389.1
XM_011540088.2 Intron XP_011538390.1

View Full Product Details