Product Details

SNP ID
rs1126766
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32274415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAACTGCCATTATCCCATAGTCC[C/G]ACTGGATGGCAAGGGCACGGTAAGA
Phenotype
MIM: 153390
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LCK PubMed Links

Gene Details

Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042771.2 205 Missense Mutation CCA,CGA P29R NP_001036236.1
NM_005356.4 205 Missense Mutation CCA,CGA P29R NP_005347.3
XM_011541453.2 205 Missense Mutation CCA,CGA P29R XP_011539755.1
XM_011541454.2 205 Missense Mutation CCA,CGA P29R XP_011539756.1
XM_011541455.2 205 Missense Mutation CCA,CGA P29R XP_011539757.1

View Full Product Details