Product Details
- SNP ID
-
rs7077285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:42436434 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCTGAGAGAAGAAGGCTTCACAC[A/G]ATCAAATTACTCTGATCTACGGGAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCNYL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs550595424] are located under a probe and SNP(s) [rs144268817] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCNYL2
- Gene Name
- cyclin Y-like 2 (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details