Product Details
- SNP ID
-
hCV64235072
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:417959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACAGACAGGCTTAATTTTACTGCA[A/G]AAACGGGTTGGCTGAAGGGAACAGG
- Phenotype
-
MIM: 605478
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANO9
PubMed Links
Gene Details
- Gene
- ANO9
- Gene Name
- anoctamin 9
There are no transcripts associated with this gene.
- Gene
- SIGIRR
- Gene Name
- single Ig and TIR domain containing
There are no transcripts associated with this gene.
View Full Product Details