Product Details

SNP ID
rs28364668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:705297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCGCCCTGTGCCCCTTCACCTC[C/G]TCCTCGTCCAGTTGACTGCTGTGGC
Phenotype
MIM: 602635 MIM: 614988
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEAF1 PubMed Links

Gene Details

Gene
DEAF1
Gene Name
DEAF1, transcription factor
There are no transcripts associated with this gene.

Gene
EPS8L2
Gene Name
EPS8 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022772.3 81 Intron NP_073609.2
XM_017018130.1 81 UTR 5 XP_016873619.1
XM_017018131.1 81 Intron XP_016873620.1
XM_017018132.1 81 Intron XP_016873621.1
Gene
TMEM80
Gene Name
transmembrane protein 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042463.2 81 Intron NP_001035928.2
NM_001276253.1 81 Intron NP_001263182.1
NM_001276274.1 81 Intron NP_001263203.1
NM_174940.3 81 Intron NP_777600.3
XM_006718206.3 81 Intron XP_006718269.3
XM_017017600.1 81 Intron XP_016873089.1

View Full Product Details