Product Details

SNP ID
rs28379116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1882979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTACATTAATAAGGTTGTATCA[C/T]CATCATCATTGTCTATTTCCAGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00254 PubMed Links

Gene Details

Gene
LINC00254
Gene Name
long intergenic non-protein coding RNA 254
There are no transcripts associated with this gene.

View Full Product Details