Product Details

SNP ID
rs28685661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37610648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGCATTTTAAAAAATCCTCAGT[A/C]TCCTTAGGAGTTACAATTGGCATCT
Phenotype
MIM: 612500 MIM: 607291
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX52 PubMed Links

Gene Details

Gene
DDX52
Gene Name
DEAD-box helicase 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291476.1 2990 UTR 3 NP_001278405.1
NM_007010.4 2990 UTR 3 NP_008941.3
XM_011524232.2 2990 UTR 3 XP_011522534.1
XM_011524233.2 2990 UTR 3 XP_011522535.1
Gene
MIR378J
Gene Name
microRNA 378j
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163544.2 2990 Intron NP_001157016.1
NM_001163545.2 2990 Intron NP_001157017.1
NM_001163546.2 2990 Intron NP_001157018.1
NM_001163547.2 2990 Intron NP_001157019.1
NM_007247.5 2990 Intron NP_009178.3
NM_080550.4 2990 Intron NP_542117.3
NM_198882.2 2990 Intron NP_942583.1
XM_005256980.4 2990 Intron XP_005257037.1
XM_017024076.1 2990 Intron XP_016879565.1
XM_017024077.1 2990 Intron XP_016879566.1
XM_017024078.1 2990 Intron XP_016879567.1
XM_017024079.1 2990 Intron XP_016879568.1
XM_017024080.1 2990 UTR 5 XP_016879569.1
XM_017024081.1 2990 UTR 5 XP_016879570.1
XM_017024082.1 2990 Intron XP_016879571.1
XM_017024083.1 2990 Intron XP_016879572.1
XM_017024084.1 2990 Intron XP_016879573.1
XM_017024085.1 2990 Intron XP_016879574.1
XM_017024086.1 2990 Intron XP_016879575.1
XM_017024087.1 2990 Intron XP_016879576.1
XM_017024088.1 2990 Intron XP_016879577.1
XM_017024089.1 2990 Intron XP_016879578.1
XM_017024090.1 2990 Intron XP_016879579.1
XM_017024091.1 2990 Intron XP_016879580.1
XM_017024092.1 2990 Intron XP_016879581.1
XM_017024093.1 2990 Intron XP_016879582.1
XM_017024094.1 2990 Intron XP_016879583.1
XM_017024095.1 2990 Intron XP_016879584.1
XM_017024096.1 2990 Intron XP_016879585.1
XM_017024097.1 2990 Intron XP_016879586.1
XM_017024098.1 2990 Intron XP_016879587.1
XM_017024099.1 2990 Intron XP_016879588.1
XM_017024100.1 2990 Intron XP_016879589.1
XM_017024101.1 2990 Intron XP_016879590.1
XM_017024102.1 2990 Intron XP_016879591.1
XM_017024103.1 2990 Intron XP_016879592.1
XM_017024104.1 2990 Intron XP_016879593.1
XM_017024105.1 2990 Intron XP_016879594.1
XM_017024106.1 2990 Intron XP_016879595.1

View Full Product Details