Product Details

SNP ID
hCV64435426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48924080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGGGGAAGGTAAATGCGTTAA[A/G]GACCACCAGCAGCCATGACCTTCTT
Phenotype
MIM: 300713 MIM: 300295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD5 PubMed Links

Gene Details

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
There are no transcripts associated with this gene.

Gene
PIM2
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
There are no transcripts associated with this gene.

View Full Product Details