Product Details

SNP ID
rs5963828
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40655783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAGAAGCCAAATGTTGAGACTA[G/T]TAGCAAAAGAACACTGTTGCCCCAA
Phenotype
MIM: 300182
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CXorf38 PubMed Links

Gene Details

Gene
CXorf38
Gene Name
chromosome X open reading frame 38
There are no transcripts associated with this gene.

Gene
MED14
Gene Name
mediator complex subunit 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004229.3 Intron NP_004220.2

View Full Product Details