Product Details

SNP ID
rs78523210
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:182409596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGCTATATTATAGGGCAGCATA[G/T]GTAGAGAACATTTTTATCCCTGCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00272 PubMed Links

Gene Details

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

Gene
TEDDM1
Gene Name
transmembrane epididymal protein 1
There are no transcripts associated with this gene.

View Full Product Details