Product Details
- SNP ID
-
rs61739738
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
28
- Location
-
Chr.1:154323118 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGAGCCACCTATGTTCTCTACCA[C/T]GGTGACAGAGGGAACAGAGGGAGCT
- Phenotype
-
MIM: 606578
MIM: 605867
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AQP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6685323] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AQP10
- Gene Name
- aquaporin 10
- Gene
- ATP8B2
- Gene Name
- ATPase phospholipid transporting 8B2
There are no transcripts associated with this gene.
View Full Product Details