Product Details
- SNP ID
-
rs61740979
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:228425233 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGCTTCTTCACGCCGCCAGTGGC[A/G]GGTGCGCTCTTGCGAGCCACCTTGG
- Phenotype
-
MIM: 602820
MIM: 606123
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST3H3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2230656] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST3H3
- Gene Name
- histone cluster 3, H3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003493.2 |
93 |
Silent Mutation |
CCC,CCT |
P31P |
NP_003484.1 |
- Gene
- TRIM17
- Gene Name
- tripartite motif containing 17
There are no transcripts associated with this gene.
View Full Product Details