Product Details
- SNP ID
-
rs72798731
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32290268 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAATTAGCTGTGAAACCTTTTGTA[C/T]GTTTTTTCACTTCTTTCATCTTTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YIPF4
PubMed Links
Gene Details
- Gene
- YIPF4
- Gene Name
- Yip1 domain family member 4
View Full Product Details