Product Details
- SNP ID
-
rs61739670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:171868665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAGATGTAAGTGTATGTTCTGGA[C/G]GGAGAACGGGTCTCATTCTGGGCAC
- Phenotype
-
MIM: 605651
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBXW11
PubMed Links
Gene Details
- Gene
- FBXW11
- Gene Name
- F-box and WD repeat domain containing 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012300.2 |
1528 |
Silent Mutation |
CCC,CCG |
P533P |
NP_036432.2 |
NM_033644.2 |
1528 |
Silent Mutation |
CCC,CCG |
P520P |
NP_387448.2 |
NM_033645.2 |
1528 |
Silent Mutation |
CCC,CCG |
P499P |
NP_387449.2 |
XM_005265855.4 |
1528 |
Silent Mutation |
CCC,CCG |
P554P |
XP_005265912.1 |
XM_005265856.2 |
1528 |
Silent Mutation |
CCC,CCG |
P522P |
XP_005265913.1 |
XM_005265857.2 |
1528 |
Silent Mutation |
CCC,CCG |
P506P |
XP_005265914.1 |
XM_005265858.2 |
1528 |
Silent Mutation |
CCC,CCG |
P501P |
XP_005265915.1 |
XM_017009279.1 |
1528 |
Silent Mutation |
CCC,CCG |
P533P |
XP_016864768.1 |
XM_017009280.1 |
1528 |
Silent Mutation |
CCC,CCG |
P501P |
XP_016864769.1 |
XM_017009281.1 |
1528 |
Silent Mutation |
CCC,CCG |
P501P |
XP_016864770.1 |
View Full Product Details