Product Details

SNP ID
rs61753580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96275054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTCAGCTTCTTTCACATCCAC[A/G]CTGAATTTGTACTCCTGGTCACATC
Phenotype
MIM: 604333 MIM: 601664 MIM: 613403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIAO1 PubMed Links

Gene Details

Gene
CIAO1
Gene Name
cytosolic iron-sulfur assembly component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004804.2 7331 Intron NP_004795.1
Gene
SNRNP200
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014014.4 7331 Silent Mutation AGC,AGT S2123S NP_054733.2
XM_017003603.1 7331 Silent Mutation AGC,AGT S2123S XP_016859092.1
Gene
TMEM127
Gene Name
transmembrane protein 127
There are no transcripts associated with this gene.

View Full Product Details