Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018912.2 | 2059 | Intron | NP_061735.1 | ||
NM_031993.1 | 2059 | Intron | NP_114382.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018915.3 | 2059 | Intron | NP_061738.1 | ||
NM_032009.2 | 2059 | Intron | NP_114398.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018916.3 | 2059 | Intron | NP_061739.2 | ||
NM_032011.1 | 2059 | Intron | NP_114400.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018917.3 | 2059 | Intron | NP_061740.2 | ||
NM_032053.2 | 2059 | Intron | NP_114442.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018922.2 | 2059 | Intron | NP_061745.1 | ||
NM_032095.1 | 2059 | Intron | NP_115266.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018923.2 | 2059 | Missense Mutation | AAA,GAA | K687E | NP_061746.1 |
NM_032096.1 | 2059 | Missense Mutation | AAA,GAA | K687E | NP_115267.1 |