Product Details

SNP ID
hCV64658019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:147179144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCATGCAGGAGGATTTCTCGC[A/G]TGATGGGAATGCTCTGGAGAGGTTC
Phenotype
MIM: 613039 MIM: 603957 MIM: 602741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHD1L PubMed Links

Gene Details

Gene
CHD1L
Gene Name
chromodomain helicase DNA binding protein 1 like
There are no transcripts associated with this gene.

Gene
FMO5
Gene Name
flavin containing monooxygenase 5
There are no transcripts associated with this gene.

Gene
PDIA3P1
Gene Name
protein disulfide isomerase family A member 3 pseudogene 1
There are no transcripts associated with this gene.

Gene
PRKAB2
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 2
There are no transcripts associated with this gene.

View Full Product Details