Product Details

SNP ID
rs41315310
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:3742866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGTGGCTTCCCCCAGCAACATG[C/G]ATACGGCATCCACGTCGTGCACAGC
Phenotype
MIM: 601990
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC27 PubMed Links

Gene Details

Gene
CCDC27
Gene Name
coiled-coil domain containing 27
There are no transcripts associated with this gene.

Gene
TP73
Gene Name
tumor protein p73
There are no transcripts associated with this gene.

Gene
TP73-AS1
Gene Name
TP73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details