Product Details

SNP ID
rs41270692
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:167053770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGGAGGTGGCCTTCCCAGGAG[A/G]GTTTGGAACATACTGGCAGCTACGG
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1696 UTR 3 NP_005805.1
XM_017000005.1 1696 UTR 3 XP_016855494.1

View Full Product Details