Product Details

SNP ID
rs41270694
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:167054244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAATGTCTGGGATGGAGGGACA[A/G]GAGAACAGGGCTTAGAAAGGAGAAG
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1222 UTR 3 NP_005805.1
XM_017000005.1 1222 UTR 3 XP_016855494.1

View Full Product Details