Product Details

SNP ID
rs41267184
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:178514220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCACTGAGAGTTCTTGCTCTCA[C/T]AGAACTTCACACTGTAACCAATCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX35 PubMed Links

Gene Details

Gene
TEX35
Gene Name
testis expressed 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170722.1 Intron NP_001164193.1
NM_001170723.1 Intron NP_001164194.1
NM_001170724.1 Intron NP_001164195.1
NM_032126.4 Intron NP_115502.2

View Full Product Details