Product Details

SNP ID
rs41304030
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:242088536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCGAGGTAGGTGAGTTATTAC[C/T]GTCTACATTACCAAGAGAAGCCCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLD5 PubMed Links

Gene Details

Gene
PLD5
Gene Name
phospholipase D family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195811.1 3156 UTR 3 NP_001182740.1
NM_001195812.1 3156 UTR 3 NP_001182741.1
NM_001320272.1 3156 UTR 3 NP_001307201.1
NM_152666.2 3156 UTR 3 NP_689879.2
XM_006711752.2 3156 UTR 3 XP_006711815.1
XM_011544115.2 3156 UTR 3 XP_011542417.1
XM_011544116.2 3156 UTR 3 XP_011542418.1
XM_011544119.2 3156 UTR 3 XP_011542421.2
XM_011544120.2 3156 UTR 3 XP_011542422.1
XM_011544121.2 3156 UTR 3 XP_011542423.1
XM_011544122.2 3156 UTR 3 XP_011542424.1
XM_017000567.1 3156 Intron XP_016856056.1
XM_017000568.1 3156 UTR 3 XP_016856057.1
XM_017000569.1 3156 UTR 3 XP_016856058.1
XM_017000570.1 3156 UTR 3 XP_016856059.1

View Full Product Details