Product Details

SNP ID
rs41272176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36741738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGGTCCCCGGGGCTCCCAGC[G/T]CATCCAGTGTCCCTGGGAGGGTGCA
Phenotype
MIM: 604209
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPNE5 PubMed Links

Gene Details

Gene
CPNE5
Gene Name
copine 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314017.1 1582 Intron NP_001300946.1
NM_001314018.1 1582 UTR 3 NP_001300947.1
NM_001314019.1 1582 UTR 3 NP_001300948.1
NM_001314020.1 1582 UTR 3 NP_001300949.1
NM_020939.1 1582 UTR 3 NP_065990.1
XM_005249247.2 1582 UTR 3 XP_005249304.1
XM_011514768.1 1582 Intron XP_011513070.1
XM_011514769.1 1582 Intron XP_011513071.1
XM_011514770.1 1582 Intron XP_011513072.1
XM_011514771.2 1582 Intron XP_011513073.1
XM_011514772.1 1582 Intron XP_011513074.1
XM_011514773.2 1582 Intron XP_011513075.1
XM_011514775.2 1582 Intron XP_011513077.1
XM_017011139.1 1582 Intron XP_016866628.1
Gene
RAB44
Gene Name
RAB44, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details