Product Details

SNP ID
rs41478248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127815362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTTCTCCCTTCCTGCCCAGCT[C/T]AGTTCACCAGTGCTGGATCCAGGTT
Phenotype
MIM: 131195 MIM: 136510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENG PubMed Links

Gene Details

Gene
ENG
Gene Name
endoglin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000118.3 2455 UTR 3 NP_000109.1
NM_001114753.2 2455 UTR 3 NP_001108225.1
NM_001278138.1 2455 UTR 3 NP_001265067.1
Gene
FPGS
Gene Name
folylpolyglutamate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018078.2 2455 Intron NP_001018088.1
NM_001288803.1 2455 Intron NP_001275732.1
NM_004957.5 2455 Intron NP_004948.4
XM_005251864.3 2455 Intron XP_005251921.1
XM_011518439.2 2455 Intron XP_011516741.1
XM_017014565.1 2455 Intron XP_016870054.1
XM_017014566.1 2455 Intron XP_016870055.1
Gene
LOC102723566
Gene Name
uncharacterized LOC102723566
There are no transcripts associated with this gene.

View Full Product Details