Product Details

SNP ID
rs41306516
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69632727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCAGGATGCAGCCTAGAGAGG[C/T]TGATTCCTGGCTGGTGCGTCAACCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C10orf35 PubMed Links

Gene Details

Gene
C10orf35
Gene Name
chromosome 10 open reading frame 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145306.2 Intron NP_660349.1
XM_005269606.1 Intron XP_005269663.1
XM_005269608.3 Intron XP_005269665.1
XM_011539455.2 Intron XP_011537757.1
XM_017015836.1 Intron XP_016871325.1

View Full Product Details