Product Details

SNP ID
rs41282220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:27155761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGAGCCCTGCGAGCTGACTTC[C/T]TTTTGGGGCGACATCAGCGGTCGGG
Phenotype
MIM: 608221 MIM: 607472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MASTL PubMed Links

Gene Details

Gene
MASTL
Gene Name
microtubule associated serine/threonine kinase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172303.2 Intron NP_001165774.1
NM_001172304.2 Intron NP_001165775.1
NM_001320756.1 Intron NP_001307685.1
NM_001320757.1 Intron NP_001307686.1
NM_032844.4 Intron NP_116233.2
XM_005252631.3 Intron XP_005252688.1
XM_005252632.3 Intron XP_005252689.1
XM_006717519.3 Intron XP_006717582.1
XM_006717520.3 Intron XP_006717583.1
XM_011519751.2 Intron XP_011518053.1
XM_017016852.1 Intron XP_016872341.1
XM_017016853.1 Intron XP_016872342.1
XM_017016854.1 Intron XP_016872343.1
XM_017016855.1 Intron XP_016872344.1
XM_017016856.1 Intron XP_016872345.1
Gene
YME1L1
Gene Name
YME1 like 1 ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253866.1 Intron NP_001240795.1
NM_014263.3 Intron NP_055078.1
NM_139312.2 Intron NP_647473.1
XM_011519300.2 Intron XP_011517602.1

View Full Product Details