Product Details

SNP ID
rs41281672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102849088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGTAGAAACTAATTTAATCCT[A/G]TGTAATAGTGAGCTTCATGGCTTTT
Phenotype
MIM: 133530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIVM PubMed Links

Gene Details

Gene
BIVM
Gene Name
basic, immunoglobulin-like variable motif containing
There are no transcripts associated with this gene.

Gene
BIVM-ERCC5
Gene Name
BIVM-ERCC5 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204425.1 Intron NP_001191354.1
Gene
ERCC5
Gene Name
ERCC excision repair 5, endonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000123.3 Intron NP_000114.2

View Full Product Details