Product Details

SNP ID
rs41282065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74762028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGCAGCGCGAGCTTCGGCCTC[A/G]GCTCTGTACCATGAAGAAGGGCCCC
Phenotype
MIM: 604990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAT9 PubMed Links

Gene Details

Gene
NAT9
Gene Name
N-acetyltransferase 9 (putative)
There are no transcripts associated with this gene.

Gene
SLC9A3R1
Gene Name
SLC9A3 regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004252.4 693 Missense Mutation CAG,CGG Q153R NP_004243.1

View Full Product Details