Product Details
- SNP ID
-
rs45443494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45940831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATTGAAATCCCAGTGACACCCA[C/T]AGGCCAGTCGGTGCCCAGCTCCCCC
- Phenotype
-
MIM: 610827
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107985388
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139030017] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107985388
- Gene Name
- pre-mRNA-splicing factor cwc22-like
There are no transcripts associated with this gene.
- Gene
- PCIF1
- Gene Name
- PDX1 C-terminal inhibiting factor 1
- Gene
- ZNF335
- Gene Name
- zinc finger protein 335
There are no transcripts associated with this gene.
View Full Product Details