Product Details
- SNP ID
-
rs41492545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23079511 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCCTTCCCTCCTCACAGTTTGGG[A/C]AACATGCACAGGATGAAGGTAAATC
- Phenotype
-
MIM: 120577
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD93
PubMed Links
Gene Details
- Gene
- CD93
- Gene Name
- CD93 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012072.3 |
6546 |
UTR 3 |
|
|
NP_036204.2 |
View Full Product Details