Product Details

SNP ID
rs41275305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160010614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATACTCAGGAAGCCGAGTGTG[C/G]TGGCCCTCCAGTTCCTGATCCCAAA
Phenotype
MIM: 601963
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTC1 PubMed Links

Gene Details

Gene
TTC1
Gene Name
tetratricopeptide repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282500.1 206 Missense Mutation GCT,GGT A29G NP_001269429.1
NM_003314.2 206 Missense Mutation GCT,GGT A29G NP_003305.1

View Full Product Details