Product Details

SNP ID
rs45479905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6925957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAAGCTTTTGCCTGCAGTGGAT[C/T]ATCCTCAGCTTGGAGTGGCTCAGCC
Phenotype
MIM: 601157 MIM: 600471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFA4 PubMed Links

Gene Details

Gene
DEFA4
Gene Name
defensin alpha 4
There are no transcripts associated with this gene.

Gene
DEFA6
Gene Name
defensin alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001926.3 120 Missense Mutation AAT,GAT N27D NP_001917.1

View Full Product Details