Product Details

SNP ID
rs41481647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59331310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAACAGAAATCACAATAAACTTA[A/G]TTTTATAGGTGGTGAAAATTGTATT
Phenotype
MIM: 601567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMAN1 PubMed Links

Gene Details

Gene
LMAN1
Gene Name
lectin, mannose binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005570.3 Intron NP_005561.1

View Full Product Details