Product Details
- SNP ID
-
rs41499255
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:59331453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAACCAATGAATAATACAGTTTG[C/T]ACCACAACAAATATAATGAAGTGGA
- Phenotype
-
MIM: 601567
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LMAN1
PubMed Links
Gene Details
- Gene
- LMAN1
- Gene Name
- lectin, mannose binding 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005570.3 |
1493 |
Silent Mutation |
GTA,GTG |
V487V |
NP_005561.1 |
View Full Product Details