Product Details
- SNP ID
-
rs45492194
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132728127 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATACACCCTTCCTGTAGATTTTGTT[A/G]ATAAAGATGTGAATTTCGTGTTGCT
- Phenotype
-
MIM: 606592
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VNN3
PubMed Links
Gene Details
- Gene
- VNN3
- Gene Name
- vanin 3
View Full Product Details