Product Details

SNP ID
rs41286546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70045493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTACAATGAAAAAAGAACCCCA[A/C]CCCAGCCACATGAATGTGACTCATT
Phenotype
MIM: 607991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC8A3 PubMed Links

Gene Details

Gene
SLC8A3
Gene Name
solute carrier family 8 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130417.2 1689 UTR 3 NP_001123889.1
NM_033262.4 1689 UTR 3 NP_150287.1
NM_058240.3 1689 UTR 3 NP_489479.1
NM_182932.2 1689 UTR 3 NP_891977.1
NM_182936.2 1689 UTR 3 NP_891981.1
NM_183002.2 1689 UTR 3 NP_892114.1
XM_006720240.3 1689 UTR 3 XP_006720303.1
XM_017021606.1 1689 UTR 3 XP_016877095.1
XM_017021607.1 1689 UTR 3 XP_016877096.1
XM_017021608.1 1689 UTR 3 XP_016877097.1
XM_017021609.1 1689 UTR 3 XP_016877098.1
XM_017021610.1 1689 Intron XP_016877099.1
XM_017021611.1 1689 UTR 3 XP_016877100.1

View Full Product Details