Product Details

SNP ID
rs45558131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79920193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCTGAAAGTCATTCAGAGCCAG[A/G]GGTTGTGATGGTCCCCTCTGTGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMS1P21 PubMed Links

Gene Details

Gene
BMS1P21
Gene Name
BMS1, ribosome biogenesis factor pseudogene 21
There are no transcripts associated with this gene.

Gene
MBL1P
Gene Name
mannose binding lectin 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details